siRNA Details
Gene Symbol: Vamp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108856.1 | NP_001102326.1 | 6 | 446 | ||
XM_006250184.2 | XP_006250246.1 | 6 | 533 | |||
XM_006250186.2 | XP_006250248.1 | 6 | 525 |
