siRNA Details
Gene Symbol: Tmem165 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024802.1 | NP_001019973.1 | 3 | 730 | ||
GenBank mRNA | BC097478.1 | - | 3 | 730 | 7458161 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmem165 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024802.1 | NP_001019973.1 | 3 | 730 | ||
GenBank mRNA | BC097478.1 | - | 3 | 730 | 7458161 |