siRNA Details
Gene Symbol: Plek | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025750.1 | NP_001020921.1 | 2 | 294 | ||
XM_006251520.1 | XP_006251582.1 | 2 | 288 | |||
GenBank mRNA | BC098846.1 | - | 2 | 294 | 7379453 | |
FQ225547.1 | - | 2 | 169 | |||
FQ232611.1 | - | 2 | 153 |
