siRNA Details
| Gene Symbol: Rnase9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008561.1 | NP_001008561.1 | 2 | 225 | ||
| XM_006251890.2 | XP_006251952.1 | 2 | 702 | |||
| XM_006251891.2 | XP_006251953.1 | 2 | 624 | |||
| GenBank mRNA | AY303815.1 | - | 2 | 225 | ||

Rnase9
Genomic Map