siRNA Details
| Gene Symbol: Psma8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108884.1 | NP_001102354.1 | 1, 2 | 207 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Psma8| Gene Symbol: Psma8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108884.1 | NP_001102354.1 | 1, 2 | 207 | ||