siRNA Details
| Gene Symbol: Olr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000539.1 | NP_001000539.1 | 1 | 306 | ||
| Gene Symbol: LOC100910456 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748751.2 | XP_003748799.2 | 1 | 30 | ||
More Information
Set Membership: |
|

Olr6
Genomic Map