siRNA Details
| Gene Symbol: Cabp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108907.1 | NP_001102377.1 | 1 | 214 | ||
| XM_006228328.2 | XP_006228390.1 | 1 | 232 | |||
| XM_006228329.2 | XP_006228391.1 | 1 | 234 | |||
| XM_006228330.2 | XP_006228392.1 | 1 | 233 | |||
| XM_006228331.2 | XP_006228393.1 | 1 | 233 | |||
| XM_006228332.2 | XP_006228394.1 | 1 | 233 | |||
| XM_006228333.2 | XP_006228395.1 | 1 | 233 | |||

Cabp5
Genomic Map