siRNA Details
Gene Symbol: Inpp5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108923.2 | NP_001102393.1 | 16 | 1580 | ||
XM_006230486.2 | XP_006230548.1 | 16 | 1836 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Inpp5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108923.2 | NP_001102393.1 | 16 | 1580 | ||
XM_006230486.2 | XP_006230548.1 | 16 | 1836 |