siRNA Details
| Gene Symbol: Ssh3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012217.1 | NP_001012217.1 | 14 | 1866 | ||
| GenBank mRNA | BC083600.1 | - | 14 | 1866 | 7133065 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ssh3| Gene Symbol: Ssh3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012217.1 | NP_001012217.1 | 14 | 1866 | ||
| GenBank mRNA | BC083600.1 | - | 14 | 1866 | 7133065 | |