siRNA Details
| Gene Symbol: Vom2r47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099506.1 | NP_001092976.1 | 1 | 229 | ||
| XM_006232462.1 | XP_006232524.1 | 2 | 334 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vom2r47| Gene Symbol: Vom2r47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099506.1 | NP_001092976.1 | 1 | 229 | ||
| XM_006232462.1 | XP_006232524.1 | 2 | 334 | |||
Set Membership: |
|