siRNA Details
Gene Symbol: Zdhhc12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013239.1 | NP_001013257.1 | 5 | 662 | ||
GenBank mRNA | AY886528.1 | - | 5 | 662 | ||
BC076393.1 | - | 5 | 662 | 7107692 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zdhhc12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013239.1 | NP_001013257.1 | 5 | 662 | ||
GenBank mRNA | AY886528.1 | - | 5 | 662 | ||
BC076393.1 | - | 5 | 662 | 7107692 |