siRNA Details
| Gene Symbol: Crb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135761.1 | NP_001129233.1 | 7 | 1181 | ||
| XM_006234103.2 | XP_006234165.1 | 6 | 1312 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Crb2| Gene Symbol: Crb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135761.1 | NP_001129233.1 | 7 | 1181 | ||
| XM_006234103.2 | XP_006234165.1 | 6 | 1312 | |||