siRNA Details
| Gene Symbol: RGD1560718 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008758795.1 | XP_008757017.1 | 2 | 879 | ||
| Gene Symbol: LOC103690981 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008758798.1 | XP_008757020.1 | 2 | 1228 | ||

RGD1560718
Genomic Map