siRNA Details
| Gene Symbol: Olr532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000563.1 | NP_001000563.1 | 1 | 656 | ||
| XM_008761992.1 | XP_008760214.1 | 2 | 700 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr532| Gene Symbol: Olr532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000563.1 | NP_001000563.1 | 1 | 656 | ||
| XM_008761992.1 | XP_008760214.1 | 2 | 700 | |||