siRNA Details
| Gene Symbol: Zfp106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762134.1 | XP_008760356.1 | 21 | 5607 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp106| Gene Symbol: Zfp106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762134.1 | XP_008760356.1 | 21 | 5607 | ||