siRNA Details
| Gene Symbol: Crls1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014258.1 | NP_001014280.1 | 5 | 827 | ||
| XM_006235078.2 | XP_006235140.1 | 5 | 1021 | |||
| XM_006235079.2 | XP_006235141.1 | 5 | 1010 | |||
| XM_006235080.2 | XP_006235142.1 | 5 | 510 | |||
| GenBank mRNA | BC085849.1 | - | 5 | 827 | 7189638 | |

Crls1
Genomic Map