siRNA Details
| Gene Symbol: Tceanc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014260.2 | NP_001014282.1 | 4 | 492 | ||
| GenBank mRNA | BC083757.1 | - | 4 | 479 | 7192791 | |
| DY316983.1 | - | 4 | 492 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tceanc2| Gene Symbol: Tceanc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014260.2 | NP_001014282.1 | 4 | 492 | ||
| GenBank mRNA | BC083757.1 | - | 4 | 479 | 7192791 | |
| DY316983.1 | - | 4 | 492 | |||