siRNA Details
Gene Symbol: Lzic | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013241.1 | NP_001013259.1 | 4 | 540 | ||
XM_006239434.2 | XP_006239496.1 | 4 | 682 | |||
XM_006239435.1 | XP_006239497.1 | 4 | 284 | |||
GenBank mRNA | BC087098.1 | - | 4 | 540 | 7191966 |
