siRNA Details
| Gene Symbol: Znf740 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006242455.2 | XP_006242517.1 | 5 | 943 | ||
| XM_006242458.2 | XP_006242520.1 | 5 | 704 | |||
| XM_008765870.1 | XP_008764092.1 | 5 | 740 | |||
| XM_008765871.1 | XP_008764093.1 | 5 | 907 | |||
| XM_008765872.1 | XP_008764094.1 | 5 | 1165 | |||
| XM_008765873.1 | XP_008764095.1 | 6 | 1021 | |||
| XM_008765874.1 | XP_008764096.1 | 6 | 818 | |||
| XM_008765875.1 | XP_008764097.1 | 4 | 590 | |||

Znf740
Genomic Map