siRNA Details
Gene Symbol: Znf740 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006242455.2 | XP_006242517.1 | 6 | 1064 | ||
XM_006242458.2 | XP_006242520.1 | 6 | 825 | |||
XM_008765870.1 | XP_008764092.1 | 6 | 861 | |||
XM_008765871.1 | XP_008764093.1 | 6 | 1028 | |||
XM_008765872.1 | XP_008764094.1 | 6 | 1286 | |||
XM_008765873.1 | XP_008764095.1 | 7 | 1142 | |||
XM_008765874.1 | XP_008764096.1 | 7 | 939 | |||
XM_008765875.1 | XP_008764097.1 | 5 | 711 |
