siRNA Details
| Gene Symbol: Smad6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109002.2 | NP_001102472.1 | 1 | 371 | ||
| XM_006243241.2 | XP_006243303.1 | 1 | 1482 | |||
| XM_006243242.2 | XP_006243304.1 | 2 | 1399 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smad6| Gene Symbol: Smad6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109002.2 | NP_001102472.1 | 1 | 371 | ||
| XM_006243241.2 | XP_006243303.1 | 1 | 1482 | |||
| XM_006243242.2 | XP_006243304.1 | 2 | 1399 | |||