siRNA Details
| Gene Symbol: Ankrd39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135014.1 | NP_001128486.1 | 2 | 176 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ankrd39| Gene Symbol: Ankrd39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135014.1 | NP_001128486.1 | 2 | 176 | ||