siRNA Details
| Gene Symbol: Slco6d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014270.1 | NP_001014292.1 | 358 | |||
| XM_006245580.2 | XP_006245642.1 | 521 | ||||
| GenBank mRNA | BC083829.1 | - | 358 | 7112354 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slco6d1| Gene Symbol: Slco6d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014270.1 | NP_001014292.1 | 358 | |||
| XM_006245580.2 | XP_006245642.1 | 521 | ||||
| GenBank mRNA | BC083829.1 | - | 358 | 7112354 | ||