siRNA Details
| Gene Symbol: Btk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007798.1 | NP_001007799.1 | 9 | 917 | ||
| GenBank mRNA | AY787790.1 | - | 9 | 917 | ||
| FQ215667.1 | - | 1061 | ||||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_013482, L08967, L10627, L29788.

Btk
Genomic Map