siRNA Details
| Gene Symbol: Prkag2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_184051.1 | NP_908940.1 | 2, 3 | 245 | ||
| XM_006235949.1 | XP_006236011.1 | 6, 7 | 913 | |||
| XM_006235950.1 | XP_006236012.1 | 6, 7 | 910 | |||
| XM_006235951.1 | XP_006236013.1 | 6, 7 | 913 | |||
| XM_006235952.1 | XP_006236014.1 | 6, 7 | 1816 | |||
| XM_006235953.2 | XP_006236015.1 | 6, 7 | 3522 | |||
| XM_006235955.2 | XP_006236017.1 | 4, 5 | 1786 | |||
| XM_006235957.2 | XP_006236019.1 | 2, 3 | 4549 | |||
| XM_008762653.1 | XP_008760875.1 | 3, 4 | 797 | |||
| XM_008762654.1 | XP_008760876.1 | 2, 3 | 4545 | |||
| GenBank mRNA | AY348865.1 | - | 2, 3 | 245 | ||
| AY376854.1 | - | 1, 2 | 98 | |||
| BC097267.1 | - | 4, 5 | 492 | |||
| FQ223364.1 | - | 6, 7 | 1503 | |||

Prkag2
Genomic Map