siRNA Details
Gene Symbol: Vkorc1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203338.1 | NP_976083.1 | 2, 3 | 348 | ||
GenBank mRNA | AY423048.1 | - | 2, 3 | 348 |
Gene Symbol: LOC103693015 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766000.1 | XP_008764222.1 | 2, 3 | 331 |
