siRNA Details
| Gene Symbol: Olr551 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000670.1 | NP_001000670.1 | 432 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr551| Gene Symbol: Olr551 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000670.1 | NP_001000670.1 | 432 | |||