siRNA Details
| Gene Symbol: Olr16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000694.1 | NP_001000694.1 | 333 | |||
| Gene Symbol: LOC100910009 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748917.2 | XP_003748965.1 | 1 | 409 | ||
| Gene Symbol: Olr294 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748914.2 | XP_003748962.1 | 409 | |||

Olr16
Genomic Map