siRNA Details
| Gene Symbol: Olr1496 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000716.1 | NP_001000716.1 | 1 | 902 | ||
| GenBank mRNA | M64385.1 | - | 1 | 902 | ||
| Gene Symbol: LOC100909578 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750853.1 | XP_003750901.1 | 1 | 902 | ||

Olr1496
Genomic Map