siRNA Details
| Gene Symbol: Olr1291 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000797.1 | NP_001000797.1 | 1 | 409 | ||
| Gene Symbol: LOC100911064 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751484.1 | XP_003751532.1 | 1 | 409 | ||

Olr1291
Genomic Map