siRNA Details
| Gene Symbol: Olr1288 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000798.1 | NP_001000798.1 | 1 | 568 | ||
| Gene Symbol: LOC100911127 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008770725.1 | XP_008768947.1 | 1 | 568 | ||

Olr1288
Genomic Map