siRNA Details
| Gene Symbol: Olr809 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000849.1 | NP_001000849.1 | 867 | |||
| Gene Symbol: LOC103690255 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762955.1 | XP_008761177.1 | 1 | 867 | ||

Olr809
Genomic Map