siRNA Details
| Gene Symbol: Olr1118 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000883.1 | NP_001000883.1 | 1 | 631 | ||
| Gene Symbol: LOC103693021 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766027.1 | XP_008764249.1 | 2 | 394 | ||

Olr1118
Genomic Map