siRNA Details
| Gene Symbol: Olr499 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000931.1 | NP_001000931.1 | 1 | 436 | ||
| Gene Symbol: LOC100912540 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749501.2 | XP_003749549.1 | 1 | 436 | ||

Olr499
Genomic Map