siRNA Details
Gene Symbol: Olr984 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001074.1 | NP_001001074.1 | 1 | 248 |
Gene Symbol: LOC102555501 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765070.1 | XP_008763292.1 | 3 | 1088 |
