siRNA Details
Gene Symbol: Olr1225 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001083.1 | NP_001001083.1 | 1 | 859 |
Gene Symbol: LOC100911999 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766103.1 | XP_008764325.1 | 871 |
