siRNA Details
| Gene Symbol: Sftpc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017342.2 | NP_059038.1 | 4 | 410 | ||
| GenBank mRNA | BC072693.1 | - | 4 | 403 | 7099876 | |
| X14221.1 | - | 4 | 411 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sftpc| Gene Symbol: Sftpc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017342.2 | NP_059038.1 | 4 | 410 | ||
| GenBank mRNA | BC072693.1 | - | 4 | 403 | 7099876 | |
| X14221.1 | - | 4 | 411 | |||