siRNA Details
Gene Symbol: Rabep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019124.1 | NP_061997.1 | 15, 16 | 2321 | ||
XM_006246795.2 | XP_006246857.1 | 14, 15 | 2379 | |||
XM_006246796.2 | XP_006246858.1 | 13, 14 | 2262 | |||
XM_008767861.1 | XP_008766083.1 | 15, 16 | 3115 | |||
GenBank mRNA | D85844.1 | - | 15, 16 | 2321 | ||
U70777.1 | - | 15, 16 | 2468 |
