siRNA Details
| Gene Symbol: App | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019288.2 | NP_062161.1 | 3, 4 | 379 | ||
| XM_006248008.2 | XP_006248070.1 | 3, 4 | 509 | |||
| XM_006248009.2 | XP_006248071.1 | 3, 4 | 509 | |||
| XM_006248010.2 | XP_006248072.1 | 3, 4 | 509 | |||
| XM_006248011.2 | XP_006248073.1 | 3, 4 | 509 | |||
| XM_006248012.2 | XP_006248074.1 | 3, 4 | 509 | |||
| GenBank mRNA | AF513015.1 | - | 3, 4 | 380 | ||
| BC062082.1 | - | 3, 4 | 492 | 5599880 | ||
| FQ221425.1 | - | 3, 4 | 512 | |||

App
Genomic Map