siRNA Details
| Gene Symbol: Cd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019295.1 | NP_062168.1 | 5, 6 | 881 | ||
| GenBank mRNA | BC098918.1 | - | 6, 7 | 771 | 7384975 | |
| D10728.1 | - | 5, 6 | 881 | |||
| X78985.1 | - | 5, 6 | 828 | |||
| Gene Symbol: LOC100911215 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006231081.2 | XP_006231143.2 | 702 | |||

Cd5
Genomic Map