siRNA Details
| Gene Symbol: Cdk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019296.1 | NP_062169.1 | 5 | 558 | ||
| XM_006256353.2 | XP_006256415.1 | 6 | 685 | |||
| GenBank mRNA | BC091549.1 | - | 6 | 643 | 7388690 | |
| FQ219810.1 | - | 5 | 561 | |||
| FQ227059.1 | - | 5 | 561 | |||
| X60767.1 | - | 5 | 558 | |||

Cdk1
Genomic Map