siRNA Details
Gene Symbol: Nr4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019328.3 | NP_062201.2 | 2 | 893 | ||
XM_006234201.2 | XP_006234263.1 | 3 | 1111 | |||
XM_008761861.1 | XP_008760083.1 | 2 | 1027 | |||
GenBank mRNA | DQ092629.1 | - | 2 | 824 | ||
DQ092630.1 | - | 2 | 660 | |||
DQ092631.1 | - | 2 | 838 | |||
DQ092632.1 | - | 2 | 828 | |||
DQ092633.1 | - | 2 | 660 | |||
DQ092634.1 | - | 2 | 660 | |||
FQ222109.1 | - | 3 | 1132 | |||
L08595.1 | - | 2 | 893 | |||
U01146.1 | - | 2 | 824 | |||
U72345.1 | - | 3 | 1117 |
