siRNA Details
| Gene Symbol: Cntn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019329.2 | NP_062202.1 | 22 | 3109 | ||
| XM_006236968.2 | XP_006237030.1 | 23 | 3297 | |||
| GenBank mRNA | U11031.1 | - | 22 | 3110 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cntn3| Gene Symbol: Cntn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019329.2 | NP_062202.1 | 22 | 3109 | ||
| XM_006236968.2 | XP_006237030.1 | 23 | 3297 | |||
| GenBank mRNA | U11031.1 | - | 22 | 3110 | ||