siRNA Details
| Gene Symbol: Rgs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019336.1 | NP_062209.1 | 2 | 239 | ||
| XM_006249974.2 | XP_006250036.1 | 2 | 423 | |||
| GenBank mRNA | BC098681.1 | - | 2 | 239 | 7385088 | |
| FQ234088.1 | - | 2 | 137 | |||

Rgs1
Genomic Map