siRNA Details
| Gene Symbol: Eif2s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019356.1 | NP_062229.1 | 6 | 668 | ||
| GenBank mRNA | BC087019.1 | - | 6 | 725 | 7191037 | |
| FQ225593.1 | - | 2 | 140 | |||
| J02646.1 | - | 6 | 668 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eif2s1| Gene Symbol: Eif2s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019356.1 | NP_062229.1 | 6 | 668 | ||
| GenBank mRNA | BC087019.1 | - | 6 | 725 | 7191037 | |
| FQ225593.1 | - | 2 | 140 | |||
| J02646.1 | - | 6 | 668 | |||