siRNA Details
| Gene Symbol: Apom | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019373.2 | NP_062246.1 | 4 | 513 | ||
| GenBank mRNA | AF207821.1 | - | 4 | 490 | ||
| BC089807.1 | - | 4 | 462 | 7323885 | ||
| FQ210093.1 | - | 4 | 512 | |||
| FQ219449.1 | - | 4 | 513 | |||

Apom
Genomic Map