siRNA Details
Gene Symbol: Nr0b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053317.1 | NP_445769.1 | 2 | 1289 | ||
GenBank mRNA | X99470.1 | - | 2 | 1289 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nr0b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053317.1 | NP_445769.1 | 2 | 1289 | ||
GenBank mRNA | X99470.1 | - | 2 | 1289 |