siRNA Details
| Gene Symbol: Nr1h2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031626.1 | NP_113814.1 | 7, 8 | 1131 | ||
| GenBank mRNA | BC157812.1 | - | 7, 8 | 1110 | ||
| U14533.1 | - | 7, 8 | 1131 | |||
| U20389.1 | - | 7, 8 | 1075 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nr1h2| Gene Symbol: Nr1h2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031626.1 | NP_113814.1 | 7, 8 | 1131 | ||
| GenBank mRNA | BC157812.1 | - | 7, 8 | 1110 | ||
| U14533.1 | - | 7, 8 | 1131 | |||
| U20389.1 | - | 7, 8 | 1075 | |||