siRNA Details
| Gene Symbol: Gpc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030828.1 | NP_110455.1 | 8 | 1608 | ||
| GenBank mRNA | BC061572.1 | - | 8 | 1606 | 6887032 | |
| L02896.1 | - | 8 | 1608 | |||
| L34067.1 | - | 8 | 1441 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpc1| Gene Symbol: Gpc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030828.1 | NP_110455.1 | 8 | 1608 | ||
| GenBank mRNA | BC061572.1 | - | 8 | 1606 | 6887032 | |
| L02896.1 | - | 8 | 1608 | |||
| L34067.1 | - | 8 | 1441 | |||