siRNA Details
Gene Symbol: Sult4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031641.1 | NP_113829.1 | 1 | 88 | ||
XM_006242114.2 | XP_006242176.1 | 1 | 880 | |||
GenBank mRNA | AF188699.1 | - | 1 | 88 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sult4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031641.1 | NP_113829.1 | 1 | 88 | ||
XM_006242114.2 | XP_006242176.1 | 1 | 880 | |||
GenBank mRNA | AF188699.1 | - | 1 | 88 |